Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270974.2 | NP_001257903.1 | 66 - 67 | 11501 | 74 |
XM_006721206.2 | XP_006721269.1 | 66 - 67 | 11395 | 74 | |
XM_011523146.2 | XP_011521448.1 | 67 - 68 | 11530 | 74 | |
XM_011523147.1 | XP_011521449.1 | 67 - 68 | 11497 | 74 | |
XM_011523148.1 | XP_011521450.1 | 67 - 68 | 11570 | 74 | |
XM_011523151.1 | XP_011521453.1 | 66 - 67 | 11427 | 74 | |
XM_011523152.1 | XP_011521454.1 | 29 - 30 | 5254 | 74 | |
XM_011523155.2 | XP_011521457.1 | 22 - 23 | 4255 | 74 | |
XM_017023346.1 | XP_016878835.1 | 66 - 67 | 11673 | 74 | |
XM_017023347.1 | XP_016878836.1 | 54 - 55 | 9912 | 74 | |
XM_017023348.1 | XP_016878837.1 | 54 - 55 | 9652 | 74 |