Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270974.2 | NP_001257903.1 | 74 - 75 | 12832 | 101 |
XM_006721206.2 | XP_006721269.1 | 74 - 75 | 12726 | 101 | |
XM_011523146.2 | XP_011521448.1 | 75 - 76 | 12861 | 101 | |
XM_011523147.1 | XP_011521449.1 | 75 - 76 | 12828 | 101 | |
XM_011523148.1 | XP_011521450.1 | 75 - 76 | 12901 | 101 | |
XM_011523151.1 | XP_011521453.1 | 74 - 75 | 12758 | 101 | |
XM_011523152.1 | XP_011521454.1 | 37 - 38 | 6585 | 101 | |
XM_011523155.2 | XP_011521457.1 | 30 - 31 | 5586 | 101 | |
XM_017023346.1 | XP_016878835.1 | 74 - 75 | 13004 | 101 | |
XM_017023347.1 | XP_016878836.1 | 62 - 63 | 11243 | 101 | |
XM_017023348.1 | XP_016878837.1 | 62 - 63 | 10983 | 101 | |
GenBank mRNA | AL137259.1 | - | 2 - 3 | 228 | 101 |
BC043273.2 | - | 9 - 10 | 1552 | 101 |