Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270974.2 | NP_001257903.1 | 80 - 81 | 14090 | 67 |
XM_006721206.2 | XP_006721269.1 | 80 - 81 | 13984 | 67 | |
XM_011523146.2 | XP_011521448.1 | 81 - 82 | 14119 | 67 | |
XM_011523147.1 | XP_011521449.1 | 81 - 82 | 14086 | 67 | |
XM_011523148.1 | XP_011521450.1 | 81 - 82 | 14159 | 67 | |
XM_011523151.1 | XP_011521453.1 | 80 - 81 | 14016 | 67 | |
XM_011523152.1 | XP_011521454.1 | 43 - 44 | 7843 | 67 | |
XM_011523155.2 | XP_011521457.1 | 36 - 37 | 6844 | 67 | |
XM_017023346.1 | XP_016878835.1 | 80 - 81 | 14262 | 67 | |
XM_017023347.1 | XP_016878836.1 | 68 - 69 | 12501 | 67 | |
XM_017023348.1 | XP_016878837.1 | 68 - 69 | 12241 | 67 | |
GenBank mRNA | AK026688.1 | - | 1 - 2 | 185 | 67 |
AK074472.1 | - | 5 - 6 | 909 | 67 | |
AL137259.1 | - | 8 - 9 | 1486 | 67 | |
BC043273.2 | - | 15 - 16 | 2810 | 67 |