Assay Design
Gene Transcripts
- Gene Symbol
- GAB2
- Entrez Gene ID
-
9846
- Gene Name
- GRB2 associated binding protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 78215290 - 78417822 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
DA674753.1 |
- |
1 - 1 |
371 |
95 |
View Full Product Details