Assay Design



Gene Transcripts

Gene Symbol
MAJIN
Entrez Gene ID
283129
Gene Name
membrane anchored junction protein
Gene Aliases
C11orf85
Chromosome Location
Chr.11: 64938230 - 64972104 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300803.1 NP_001287732.1 4 - 5 425 146
GenBank mRNA BC033501.1 - 4 - 5 357 146

View Full Product Details