Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300803.1 | NP_001287732.1 | 6 - 7 | 580 | 79 |
NM_001318808.1 | NP_001305737.1 | 9 - 10 | 1528 | 79 | |
XM_005273918.2 | XP_005273975.1 | 9 - 10 | 808 | 79 | |
XM_006718502.2 | XP_006718565.1 | 9 - 10 | 925 | 79 | |
XM_006718503.3 | XP_006718566.1 | 9 - 10 | 811 | 79 | |
XM_006718505.2 | XP_006718568.1 | 6 - 7 | 691 | 79 | |
XM_011544940.2 | XP_011543242.1 | 9 - 10 | 806 | 79 | |
XM_011544941.2 | XP_011543243.1 | 3 - 4 | 507 | 79 | |
GenBank mRNA | AK093130.1 | - | 9 - 10 | 1522 | 79 |
BC033501.1 | - | 6 - 7 | 512 | 79 | |
LC068587.1 | - | 7 - 8 | 652 | 79 |