Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301020.1 | NP_001287949.1 | 3 - 4 | 571 | 174 |
NM_004152.3 | NP_004143.1 | 3 - 4 | 577 | 174 | |
GenBank mRNA | AF090094.1 | - | - | 1504 | 174 |
AK307221.1 | - | 3 - 4 | 543 | 174 | |
AL542477.3 | - | 1 - 2 | 148 | 173 | |
BC010063.1 | - | 3 - 4 | 527 | 174 | |
BI667301.1 | - | 3 - 4 | 548 | 174 | |
BM423450.1 | - | 3 - 4 | 526 | 174 | |
D78361.1 | - | 3 - 4 | 608 | 174 | |
D87914.1 | - | 3 - 4 | 524 | 174 | |
U09202.1 | - | 3 - 4 | 509 | 174 |