Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301073.1 | NP_001288002.1 | 4 - 5 | 793 | 115 |
NM_001301075.1 | NP_001288004.1 | 3 - 4 | 647 | 115 | |
NM_001301077.1 | NP_001288006.1 | 4 - 5 | 705 | 115 | |
NM_001301080.1 | NP_001288009.1 | 3 - 4 | 604 | 115 | |
NM_001301082.1 | NP_001288011.1 | 3 - 4 | 317 | 115 | |
NM_001301083.1 | NP_001288012.1 | 2 - 3 | 437 | 115 | |
NM_004779.5 | NP_004770.4 | 4 - 5 | 841 | 115 | |
XM_011537706.1 | XP_011536008.1 | 5 - 6 | 815 | 115 | |
XM_011537708.2 | XP_011536010.1 | 3 - 4 | 318 | 115 | |
XM_017010048.1 | XP_016865537.1 | 5 - 6 | 700 | 115 | |
XM_017010049.1 | XP_016865538.1 | 5 - 6 | 1295 | 115 | |
XM_017010050.1 | XP_016865539.1 | 5 - 6 | 1280 | 115 | |
XM_017010051.1 | XP_016865540.1 | 5 - 6 | 1330 | 115 | |
XM_017010057.1 | XP_016865546.1 | 5 - 6 | 858 | 115 | |
GenBank mRNA | AF053318.1 | - | 4 - 5 | 715 | 115 |
AF087844.1 | - | 3 - 4 | 543 | 115 | |
AF180476.1 | - | 3 - 4 | 543 | 115 | |
AK025358.1 | - | 4 - 5 | 762 | 115 | |
AK297381.1 | - | 3 - 4 | 512 | 115 | |
AK299589.1 | - | 5 - 6 | 673 | 115 | |
AK309359.1 | - | 4 - 5 | 595 | 115 | |
AK314317.1 | - | 4 - 5 | 793 | 115 | |
AK315864.1 | - | 3 - 4 | 496 | 115 | |
AK316221.1 | - | 3 - 4 | 317 | 115 | |
AK316269.1 | - | 2 - 3 | 435 | 115 | |
AL122045.1 | - | 4 - 5 | 722 | 115 | |
BC017366.2 | - | 4 - 5 | 652 | 115 | |
BT006857.1 | - | 3 - 4 | 471 | 115 | |
BX647124.1 | - | 2 - 3 | 184 | 115 |