Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003239.3 | NP_003230.1 | 2 - 3 | 1645 | 63 |
GenBank mRNA | AK313564.1 | - | 2 - 3 | 726 | 63 |
BC018503.1 | - | 2 - 3 | 616 | 63 | |
BT007287.1 | - | 2 - 3 | 516 | 63 | |
EF560714.1 | - | 2 - 3 | 629 | 63 | |
J03241.1 | - | 2 - 3 | 778 | 63 | |
X14149.1 | - | 2 - 3 | 769 | 63 |