Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300747.1 | NP_001287676.1 | 1 - 2 | 274 | 104 |
NM_001300748.1 | NP_001287677.1 | 1 - 2 | 274 | 104 | |
NM_024587.3 | NP_078863.2 | 1 - 2 | 274 | 104 | |
GenBank mRNA | AK026006.1 | - | 1 - 2 | 81 | 104 |
AK225405.1 | - | 1 - 2 | 81 | 104 | |
BC064520.1 | - | 1 - 2 | 93 | 104 | |
CR457348.1 | - | 1 - 2 | 57 | 104 | |
HY029536.1 | - | 1 - 2 | 108 | 104 | |
HY108001.1 | - | 1 - 2 | 276 | 104 |