Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024587.3 | NP_078863.2 | 2 - 3 | 399 | 108 |
GenBank mRNA | AK026006.1 | - | 2 - 3 | 206 | 108 |
AK225405.1 | - | 2 - 3 | 206 | 108 | |
BC064520.1 | - | 2 - 3 | 218 | 108 | |
CR457348.1 | - | 2 - 3 | 182 | 108 |