Assay Design



Gene Transcripts

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
Chromosome Location
Chr.3: 151085665 - 151436677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053002.5 NP_443728.3 18 - 19 2852 103
XM_006713487.3 XP_006713550.1 20 - 21 3229 103
XM_011512390.2 XP_011510692.1 19 - 20 3124 103
XM_011512394.2 XP_011510696.1 20 - 21 3229 103
XM_011512399.2 XP_011510701.1 20 - 21 3229 103
XM_017005676.1 XP_016861165.1 20 - 21 3229 103
XM_017005677.1 XP_016861166.1 20 - 21 3229 103
XM_017005678.1 XP_016861167.1 20 - 21 3229 103
XM_017005679.1 XP_016861168.1 19 - 20 2959 103
XM_017005680.1 XP_016861169.1 18 - 19 2947 103
XM_017005681.1 XP_016861170.1 5 - 6 615 103
GenBank mRNA AB046855.1 - 5 - 6 595 103
AF399708.1 - 18 - 19 2761 103
AK022714.1 - 2 - 3 304 103
BC144559.1 - 16 - 17 2336 103

View Full Product Details