Assay Design



Gene Transcripts

Gene Symbol
APOBEC3H
Entrez Gene ID
164668
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3H
Gene Aliases
A3H, ARP-10, ARP10
Chromosome Location
Chr.22: 39097204 - 39104067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181773.4 NP_861438.3 4 - 5 670 103
GenBank mRNA BQ052182.1 - 4 - 5 609 103

View Full Product Details