Assay Design



Gene Transcripts

Gene Symbol
TSEN15
Entrez Gene ID
116461
Gene Name
tRNA splicing endonuclease subunit 15
Gene Aliases
C1orf19, PCH2F, sen15
Chromosome Location
Chr.1: 184050207 - 184095846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127394.3 NP_001120866.1 3 - 4 455 70
GenBank mRNA AK296655.1 - 3 - 4 350 70
BM887891.1 - 3 - 4 287 70

View Full Product Details