Assay Design



Gene Transcripts

Gene Symbol
CEP57L1
Entrez Gene ID
285753
Gene Name
centrosomal protein 57 like 1
Gene Aliases
C6orf182, bA487F23.2, cep57R
Chromosome Location
Chr.6: 109095130 - 109167696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083535.1 NP_001077004.1 8 - 9 1232 104
NM_001271852.1 NP_001258781.1 6 - 7 1081 104
NM_001271853.1 NP_001258782.1 6 - 7 1081 104
NM_173830.4 NP_776191.1 6 - 7 847 104
XM_005266935.4 XP_005266992.3 6 - 7 780 104
XM_005266936.4 XP_005266993.3 6 - 7 780 104
XM_005266937.4 XP_005266994.3 6 - 7 732 104
XM_005266938.4 XP_005266995.3 6 - 7 780 104
XM_006715462.3 XP_006715525.1 7 - 8 1144 104
XM_011535759.2 XP_011534061.1 6 - 7 1094 104
XM_011535762.2 XP_011534064.1 4 - 5 368 104
XM_011535763.2 XP_011534065.2 6 - 7 780 104
XM_017010764.1 XP_016866253.1 7 - 8 1283 104
XM_017010765.1 XP_016866254.1 8 - 9 1538 104
XM_017010767.1 XP_016866256.1 6 - 7 1094 104
XM_017010768.1 XP_016866257.1 6 - 7 908 104
XM_017010769.1 XP_016866258.1 6 - 7 847 104
XM_017010770.1 XP_016866259.1 8 - 9 1538 104
XM_017010771.1 XP_016866260.1 4 - 5 373 104
XM_017010772.1 XP_016866261.1 5 - 6 883 104
GenBank mRNA AK289910.1 - 6 - 7 713 104
BC033448.2 - 6 - 7 847 104
BC064365.1 - 6 - 7 1076 104
DB204503.1 - 4 - 5 453 104

View Full Product Details