Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297559.1 | NP_001284488.1 | 3 - 4 | 914 | 117 |
NM_053044.4 | NP_444272.1 | 3 - 4 | 914 | 117 | |
XM_011513596.2 | XP_011511898.1 | 3 - 4 | 922 | 117 | |
GenBank mRNA | AL547377.3 | - | 3 - 4 | 917 | 118 |
AY040094.2 | - | 3 - 4 | 931 | 117 | |
AY280665.1 | - | 3 - 4 | 902 | 117 | |
AY280666.1 | - | 3 - 4 | 817 | 117 | |
BC034390.1 | - | 3 - 4 | 914 | 117 | |
BC035717.1 | - | 3 - 4 | 914 | 117 |