Assay Design



Gene Transcripts

Gene Symbol
UGT2B4
Entrez Gene ID
7363
Gene Name
UDP glucuronosyltransferase family 2 member B4
Gene Aliases
HLUG25, UDPGT2B4, UDPGTH1, UDPGTh-1, UGT2B11
Chromosome Location
Chr.4: 69480165 - 69526014 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297616.1 NP_001284545.1 1 - 2 326 101
GenBank mRNA AK300084.1 - 1 - 2 326 101

View Full Product Details