Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104236.1 | 3 - 4 | 333 | 96 | |
GenBank mRNA | BX352732.2 | - | 3 - 4 | 188 | 96 |
BX647991.1 | - | 3 - 4 | 265 | 96 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | melanin concentrating hormone receptor 1 | MCHR1 | GPR24, MCH-1R, MCH1R, SLC-1, SLC1 | -96.2kb | Anti-sense | Chr. - 40679178 - 40682814 on Build GRCh38 |
Downstream | microRNA 4766 | MIR4766 | mir-4766 | 19.3kb | Sense | Chr. - 40813883 - 40813958 on Build GRCh38 |