Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104447.1 | 1 - 2 | 187 | 99 | |
GenBank mRNA | BQ777217.1 | - | 1 - 2 | 94 | 99 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | unc-45 myosin chaperone A | UNC45A | GC-UNC45, GCUNC-45, GCUNC45, IRO039700, SMAP-1, SMAP1, UNC-45A | Anti-sense | Chr. - 90929980 - 90954093 on Build GRCh38 | |
Downstream | RCC1 domain containing 1 | RCCD1 | 22.1kb | Anti-sense | Chr. - 90954864 - 90964858 on Build GRCh38 |