Assay Design
Gene Transcripts
- Gene Symbol
- CFAP47
- Entrez Gene ID
-
286464
- Gene Name
- cilia and flagella associated protein 47
- Gene Aliases
-
CHDC2, CXorf22, CXorf30, CXorf59
- Chromosome Location
-
Chr.X: 35919734 - 36385319 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MAGE family member B16 |
MAGEB16 |
|
-189.4kb
|
Sense |
Chr. - 35798317 - 35803753 on Build GRCh38 |
Downstream |
uncharacterized LOC101928627 |
LOC101928627 |
|
368.2kb
|
Anti-sense |
Chr. - 36365626 - 36440292 on Build GRCh38 |
View Full Product Details