Assay Design
Gene Transcripts
- Gene Symbol
- UPP1
- Entrez Gene ID
-
7378
- Gene Name
- uridine phosphorylase 1
- Gene Aliases
-
UDRPASE, UP, UPASE, UPP
- Chromosome Location
-
Chr.7: 48088308 - 48108746 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_109837.1 |
|
4 - 5 |
386 |
98 |
GenBank mRNA |
BC053592.1 |
- |
4 - 5 |
296 |
98 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BM765489.1 |
- |
4 - 5 |
440 |
|
Chr. 7 - 48089263 - 48107083 [+] on Build GRCh38 |
98 |
GenBankmRNA |
BM811584.1 |
- |
|
362 |
|
Chr. - - [-] on Build |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 7 open reading frame 57 |
C7orf57 |
|
-40.5kb
|
Sense |
Chr. - 48035511 - 48061297 on Build GRCh38 |
Downstream |
ATP binding cassette subfamily A member 13 |
ABCA13 |
|
64.4kb
|
Sense |
Chr. - 48171424 - 48647497 on Build GRCh38 |
View Full Product Details