Assay Design
Gene Transcripts
- Gene Symbol
- CRNDE
- Entrez Gene ID
-
643911
- Gene Name
- colorectal neoplasia differentially expressed (non-protein coding)
- Gene Aliases
-
CRNDEP, LINC00180, NCRNA00180, PNAS-108, lincIRX5
- Chromosome Location
-
Chr.16: 54918863 - 54929189 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_110453.1 |
|
3 - 4 |
615 |
113 |
GenBank mRNA |
DB446128.1 |
- |
3 - 4 |
238 |
113 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927480 |
LOC101927480 |
|
-75.3kb
|
Sense |
Chr. - 54847249 - 54848233 on Build GRCh38 |
Downstream |
iroquois homeobox 5 |
IRX5 |
HMMS, IRX-2a, IRXB2 |
6.5kb
|
Anti-sense |
Chr. - 54931199 - 54934485 on Build GRCh38 |
View Full Product Details