Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110844.1 | 1 - 2 | 170 | 115 | |
GenBank mRNA | AL832096.1 | - | 1 - 2 | 170 | 115 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | clavesin 2 | CLVS2 | C6orf212, C6orf213, RLBP1L2, bA160A10.4 | -366.7kb | Sense | Chr. - 122993693 - 123081282 on Build GRCh38 |
Downstream | Na+/K+ transporting ATPase interacting 2 | NKAIN2 | FAM77B, NKAIP2, TCBA, TCBA1 | 334.9kb | Sense | Chr. - 123803840 - 124825652 on Build GRCh38 |