Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120626.1 | 1 - 2 | 266 | 82 | |
GenBank mRNA | DA356569.1 | - | 1 - 2 | 241 | 82 |
DB168259.1 | - | 1 - 2 | 267 | 82 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | cilia and flagella associated protein 46 | CFAP46 | C10orf123, C10orf124, C10orf92, C10orf93, TTC40, bA288G11.4, bA288G11.5, bB137A17.2, bB137A17.3 | -804.0 | Sense | Chr. - 132808392 - 132942585 on Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1167 | LINC01167 | TCONS_00018075 | 14.0kb | Sense | Chr. - 132961340 - 132962237 on Build GRCh38 |