Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301132.1 | NP_001288061.1 | 4 - 5 | 2231 | 81 |
NM_014952.4 | NP_055767.3 | 4 - 5 | 2107 | 81 | |
NR_125372.1 | 2 - 3 | 278 | 81 | ||
XM_011521366.2 | XP_011519668.2 | 4 - 5 | 2231 | 81 | |
XM_011521367.2 | XP_011519669.2 | 4 - 5 | 2233 | 81 | |
XM_011521368.2 | XP_011519670.2 | 4 - 5 | 2230 | 81 | |
XM_011521369.2 | XP_011519671.1 | 4 - 5 | 2085 | 81 | |
GenBank mRNA | AB023162.1 | - | 4 - 5 | 2231 | 81 |
AL833923.1 | - | 3 - 4 | 1199 | 81 | |
BC022782.2 | - | 4 - 5 | 2046 | 81 | |
HY353002.1 | - | 3 - 4 | 81 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | isovaleryl-CoA dehydrogenase | IVD | ACAD2 | -27.9kb | Sense | Chr. - 40405485 - 40435948 on Build GRCh38 |
Downstream | carbohydrate sulfotransferase 14 | CHST14 | ATCS, D4ST1, EDSMC1, HNK1ST | 6.4kb | Sense | Chr. - 40470961 - 40473158 on Build GRCh38 |