Assay Design
Gene Transcripts
- Gene Symbol
- CIB2
- Entrez Gene ID
-
10518
- Gene Name
- calcium and integrin binding family member 2
- Gene Aliases
-
DFNB48, KIP2, USH1J
- Chromosome Location
-
Chr.15: 78104606 - 78131976 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125435.1 |
|
2 - 3 |
405 |
70 |
GenBank mRNA |
AI817087.1 |
- |
2 - 3 |
141 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SH2 domain containing 7 |
SH2D7 |
|
-5.2kb
|
Anti-sense |
Chr. - 78089895 - 78104051 on Build GRCh38 |
Downstream |
isocitrate dehydrogenase 3 (NAD(+)) alpha |
IDH3A |
|
25.6kb
|
Anti-sense |
Chr. - 78149326 - 78170542 on Build GRCh38 |
View Full Product Details