Assay Design
Gene Transcripts
- Gene Symbol
- ATAD2B
- Entrez Gene ID
-
54454
- Gene Name
- ATPase family, AAA domain containing 2B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 23689710 - 23927114 on Build GRCh38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
kelch like family member 29 |
KLHL29 |
KBTBD9 |
-101.7kb
|
Anti-sense |
Chr. - 23385419 - 23708613 on Build GRCh38 |
Downstream |
UBX domain protein 2A |
UBXN2A |
UBXD4 |
110.6kb
|
Anti-sense |
Chr. - 23927201 - 24001000 on Build GRCh38 |
View Full Product Details