Assay Design



Gene Transcripts

Gene Symbol
FOXC2-AS1
Entrez Gene ID
103752587
Gene Name
FOXC2 antisense RNA 1
Gene Aliases
ODRUL
Chromosome Location
Chr.16: 86565145 - 86567761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_125795.1 1 - 2 142 102
GenBank mRNA AW271272.1 - 1 - 2 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized protein FLJ30679 FLJ30679 -7.8kb Anti-sense Chr. - 86555320 - 86557299 on Build GRCh38
Downstream forkhead box L1 FOXL1 FKH6, FKHL11, FREAC7 10.7kb Anti-sense Chr. - 86578509 - 86581698 on Build GRCh38

View Full Product Details