Assay Design
Gene Transcripts
- Gene Symbol
- MFAP3L
- Entrez Gene ID
-
9848
- Gene Name
- microfibrillar associated protein 3 like
- Gene Aliases
-
NYD-sp9
- Chromosome Location
-
Chr.4: 169986597 - 170033028 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125890.1 |
|
1 - 2 |
239 |
92 |
GenBank mRNA |
BC031941.1 |
- |
1 - 2 |
242 |
92 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX112039.1 |
- |
1 - 2 |
215 |
|
Chr. 4 - 170031770 - 170033004 [-] on Build GRCh38 |
92 |
GenBankmRNA |
BG718733.1 |
- |
1 - 2 |
247 |
|
Chr. 4 - 170031770 - 170033031 [-] on Build GRCh38 |
92 |
GenBankmRNA |
BQ438469.1 |
- |
1 - 2 |
208 |
|
Chr. 4 - 170031783 - 170032996 [-] on Build GRCh38 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC100506085 |
LOC100506085 |
|
-56.2kb
|
Sense |
Chr. - 169917761 - 169975902 on Build GRCh38 |
Downstream |
aminoadipate aminotransferase |
AADAT |
KAT2, KATII, KYAT2 |
27.2kb
|
Sense |
Chr. - 170060222 - 170091699 on Build GRCh38 |
View Full Product Details