Assay Design
Gene Transcripts
- Gene Symbol
- HSP90AB4P
- Entrez Gene ID
-
664618
- Gene Name
- heat shock protein 90 alpha family class B member 4, pseudogene
- Gene Aliases
-
HSP90Bd
- Chromosome Location
-
Chr.15: 58690473 - 58693607 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073415.2 |
|
1 - 1 |
322 |
108 |
GenBank mRNA |
DB113910.1 |
- |
1 - 1 |
322 |
108 |
View Full Product Details