Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_102430.1 | 2 - 3 | 217 | 114 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BQ187924.1 | - | 193 | Chr. - - [-] on Build | 114 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | MRG/MORF4L binding protein | MRGBP | C20orf20, Eaf7, MRG15BP, URCC4 | -34.0 | Anti-sense | Chr. - 62796405 - 62801738 on Build GRCh38 |
Downstream | opioid growth factor receptor | OGFR | 2.7kb | Anti-sense | Chr. - 62804825 - 62814000 on Build GRCh38 |