Assay Design
Gene Transcripts
- Gene Symbol
- LINC01100
- Entrez Gene ID
-
730109
- Gene Name
- long intergenic non-protein coding RNA 1100
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 160020649 - 160031423 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104132.1 |
|
1 - 2 |
95 |
112 |
GenBank mRNA |
AA453488.1 |
- |
3 - 4 |
217 |
112 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
IL12A antisense RNA 1 |
IL12A-AS1 |
|
|
Anti-sense |
Chr. - 159913400 - 160207092 on Build GRCh38 |
Downstream |
uncharacterized LOC105374314 |
LOC105374314 |
|
5.4kb
|
Anti-sense |
Chr. - 160032259 - 160040364 on Build GRCh38 |
View Full Product Details