Assay Design
Gene Transcripts
- Gene Symbol
- MAFA
- Entrez Gene ID
-
389692
- Gene Name
- MAF bZIP transcription factor A
- Gene Aliases
-
RIPE3b1, hMafA
- Chromosome Location
-
Chr.8: 143428060 - 143430432 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_201589.3 |
NP_963883.2 |
1 - 1 |
1624 |
68 |
View Full Product Details