Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CHMP2B | NM_001244644.1 | NP_001231573.1 | ||
NM_014043.3 | NP_054762.2 | |||
XM_011533576.2 | XP_011531878.1 | |||
AF151842.1 | AAD34079.1 | |||
AK002165.1 | ||||
AK002180.1 | ||||
AK222654.1 | BAD96374.1 | |||
AK296072.1 | ||||
AK316585.1 | ||||
AL080122.1 | CAB45721.1 | |||
BC001553.1 | AAH01553.1 | |||
BI821848.1 | ||||
BM828410.1 | ||||
CR533456.1 | CAG38487.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953231 | Chr3:87227951 - 87252550 on Build GRCh38 | Duplication | CHMP2B |
esv3568885 | Chr3:87182571 - 87518398 on Build GRCh38 | Loss | POU1F1 MIR4795 CHMP2B |
nsv527279 | Chr3:86796784 - 87814130 on Build GRCh38 | Gain | POU1F1 VGLL3 LINC00506 MIR4795 HTR1F CHMP2B |