Product Details
- Assay Reference Genome
Location
Chr.6:43602475 on build GRCh38
- Cytoband
- 6p21.1
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs111259865] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 5429
- Gene Symbol
- POLH
- Gene Name
- DNA polymerase eta
- Gene Aliases
- RAD30, RAD30A, XP-V, XPV
- Location
- Chr.6:43576141-43620523 on build GRCh38
- Assay Gene Location
- Within Intron 6
View Full Product Details