Product Details

Assay Reference Genome
Location

Chr.6:39814810 on build GRCh38
Cytoband
6p21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
23500
Gene Symbol
DAAM2
Gene Name
dishevelled associated activator of morphogenesis 2
Gene Aliases
dJ90A20A.1
Location
Chr.6:39792366-39904877 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DAAM2 NM_001201427.1 NP_001188356.1
NM_015345.3 NP_056160.2
XM_006715043.2 XP_006715106.1
XM_006715046.3 XP_006715109.1
XM_017010630.1 XP_016866119.1
AB002379.2
AK310941.1
AL596701.1
AL833083.2 CAD89973.1

View Full Product Details