Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178328.1 | NP_847898.1 | 2 - 3 | 231 | 78 |
XM_017005685.1 | XP_016861174.1 | 2 - 3 | 1151 | 78 | |
XM_017005686.1 | XP_016861175.1 | 3 - 4 | 1332 | 78 | |
GenBank mRNA | AF262301.1 | - | 2 - 3 | 152 | 78 |