Product Details

Assay Reference Genome
Location

Chr.7:55046274 on build GRCh38
Cytoband
7p11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1956
Gene Symbol
EGFR
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
Location
Chr.7:55019032-55207338 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
EGFR NM_005228.3 NP_005219.2
NM_201282.1 NP_958439.1
NM_201283.1 NP_958440.1
NM_201284.1 NP_958441.1
AB209442.1 BAD92679.1
AF125253.1 AAG43240.1
AK225422.1
AK290352.1
AW163038.1
AY550966.1 AAT52212.1
AY698023.1 AAT97978.1
AY698024.1 AAT97979.1
BC094761.1 AAH94761.1
DQ088980.1
GU255993.1
HQ912715.1
K03193.1 AAA52371.1
U48722.1 AAC50802.1
U95089.1 AAB53063.1
X00588.1 CAA25240.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2761332 Chr7:54574291 - 55097556 on Build GRCh38 Gain LOC100996654 EGFR SEC61G LOC107986797
nsv607029 Chr7:54579095 - 55086688 on Build GRCh38 Gain LOC100996654 EGFR SEC61G LOC107986797
nsv5753 Chr7:55045323 - 55070844 on Build GRCh38 Insertion EGFR
esv2734425 Chr7:53868335 - 55917254 on Build GRCh38 Deletion LOC100996654 LANCL2 VSTM2A ELDR ZNF713 HPVC1 LINC01445 EGFR-AS1 FKBP9P1 EGFR VOPP1 PSPHP1 SEC61G LOC101060341 LOC107986797 VSTM2A-OT1 SEPT14
nsv1030676 Chr7:54583600 - 55151489 on Build GRCh38 Gain LOC100996654 EGFR SEC61G LOC107986797

View Full Product Details