Product Details
- Assay Reference Genome
Location
Chr.7:92488497 on build GRCh38
- Cytoband
- 7q21.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 57798
- Gene Symbol
- GATAD1
- Gene Name
- GATA zinc finger domain containing 1
- Gene Aliases
- CMD2B, ODAG, RG083M05.2
- Location
- Chr.7:92447448-92494631 on build GRCh38
- Assay Gene Location
Gene Symbol |
Transcript Accession |
Exon Location |
Assay Location |
Protein ID |
GATAD1 |
|
|
|
|
- Entrez Gene ID
- 5189
- Gene Symbol
- PEX1
- Gene Name
- peroxisomal biogenesis factor 1
- Gene Aliases
- HMLR1, PBD1A, PBD1B, ZWS, ZWS1
- Location
- Chr.7:92487023-92528531 on build GRCh38
- Assay Gene Location
- Within Intron 23
View Full Product Details