Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
EGFR | NM_005228.3 | NP_005219.2 | ||
NM_201282.1 | NP_958439.1 | |||
NM_201283.1 | NP_958440.1 | |||
NM_201284.1 | NP_958441.1 | |||
AB209442.1 | BAD92679.1 | |||
AF125253.1 | AAG43240.1 | |||
AK225422.1 | ||||
AK290352.1 | ||||
AW163038.1 | ||||
AY550966.1 | AAT52212.1 | |||
AY698023.1 | AAT97978.1 | |||
AY698024.1 | AAT97979.1 | |||
BC094761.1 | AAH94761.1 | |||
DQ088980.1 | ||||
GU255993.1 | ||||
HQ912715.1 | ||||
K03193.1 | AAA52371.1 | |||
U48722.1 | AAC50802.1 | |||
U95089.1 | AAB53063.1 | |||
X00588.1 | CAA25240.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761332 | Chr7:54574291 - 55097556 on Build GRCh38 | Gain | EGFR SEC61G LOC100996654 LOC107986797 |
nsv607029 | Chr7:54579095 - 55086688 on Build GRCh38 | Gain | EGFR SEC61G LOC100996654 LOC107986797 |
esv2734425 | Chr7:53868335 - 55917254 on Build GRCh38 | Deletion | EGFR-AS1 LINC01445 VSTM2A-OT1 VSTM2A VOPP1 ZNF713 ELDR LANCL2 PSPHP1 HPVC1 EGFR SEPT14 SEC61G FKBP9P1 LOC100996654 LOC107986797 LOC101060341 |
nsv528853 | Chr7:54997426 - 55020722 on Build GRCh38 | Loss | EGFR |
nsv1030676 | Chr7:54583600 - 55151489 on Build GRCh38 | Gain | EGFR SEC61G LOC100996654 LOC107986797 |
nsv5752 | Chr7:54995331 - 55040249 on Build GRCh38 | Deletion | EGFR |