Product Details

Assay Reference Genome
Location

Chr.15:68210424 on build GRCh38
Cytoband
15q23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
54982
Gene Symbol
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
Location
Chr.15:68206992-68229742 on build GRCh38
Assay Gene Location
Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CLN6 NM_017882.2 NP_060352.1
AK000568.1 BAA91260.1
AK027604.1 BAB55226.1
AK074753.1 BAC11181.1
AK291175.1
AK293155.1
AK293197.1
BC010849.1 AAH10849.1
BC013130.1 AAH13130.1
CR457244.1 CAG33525.1
KU178671.1
KU178672.1
KU178673.1
KU178674.1

View Full Product Details