Product Details

Assay Reference Genome
Location

Chr.22:26459967 on build GRCh38
Cytoband
22q12.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
89781
Gene Symbol
HPS4
Gene Name
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases
BLOC3S2, LE
Location
Chr.22:26443613-26483863 on build GRCh38
Assay Gene Location
Within Intron 14
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
HPS4 NM_022081.5 NP_071364.4
NM_152841.2 NP_690054.1
NR_073135.1
NR_073136.1
XM_006724353.3 XP_006724416.1
XM_006724354.3 XP_006724417.1
XM_006724360.3 XP_006724423.1
XM_011530485.1 XP_011528787.1
XM_011530486.2 XP_011528788.1
XM_011530487.2 XP_011528789.1
XM_011530488.2 XP_011528790.1
XM_011530489.2 XP_011528791.1
XM_011530490.2 XP_011528792.1
XM_011530491.2 XP_011528793.1
XM_011530492.1 XP_011528794.1
XM_011530493.2 XP_011528795.1
XM_011530494.2 XP_011528796.1
XM_011530495.2 XP_011528797.1
XM_011530496.2 XP_011528798.1
XM_017029042.1 XP_016884531.1
XM_017029043.1 XP_016884532.1
XM_017029044.1 XP_016884533.1
XM_017029045.1 XP_016884534.1
XM_017029046.1 XP_016884535.1
XM_017029047.1 XP_016884536.1
XM_017029048.1 XP_016884537.1
XM_017029049.1 XP_016884538.1
XM_017029050.1 XP_016884539.1
XM_017029051.1 XP_016884540.1
XM_017029052.1 XP_016884541.1
XM_017029053.1 XP_016884542.1
XM_017029054.1 XP_016884543.1
XM_017029055.1 XP_016884544.1
XM_017029056.1 XP_016884545.1
XM_017029057.1 XP_016884546.1
XM_017029058.1 XP_016884547.1
XM_017029059.1 XP_016884548.1
XM_017029060.1 XP_016884549.1
XM_017029061.1 XP_016884550.1
XM_017029062.1 XP_016884551.1
XM_017029063.1 XP_016884552.1
XM_017029064.1 XP_016884553.1
AB051454.1 BAB33337.1
AK054717.1
AK057648.1 BAB71540.1
AK095406.1
AK123691.1
AK290211.1
AL365512.1 CAB97208.1
AL713795.1 CAD28549.1
AY043416.1 AAK95330.1
BC065030.1 AAH65030.1
CR456392.1 CAG30278.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv519472 Chr22:26458475 - 26463976 on Build GRCh38 Loss HPS4

View Full Product Details