Product Details

Assay Reference Genome
Location

Chr.11:47217468 on build GRCh38
Cytoband
11p11.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1643
Gene Symbol
DDB2
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
Location
Chr.11:47214942-47239218 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DDB2 NM_000107.2 NP_000098.1
NM_001300734.1 NP_001287663.1
AB107037.1 BAD12557.1
AB107038.1 BAD12558.1
AB107039.1 BAD12559.1
AB107040.1 BAD12560.1
AK091640.1
AK313262.1
BC000093.2 AAH00093.1
BT007139.1 AAP35803.1
U18300.1 AAB07897.1

View Full Product Details