Product Details

Assay Reference Genome
Location

Chr.18:51056284 on build GRCh38
Cytoband
18q21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
4089
Gene Symbol
SMAD4
Gene Name
SMAD family member 4
Gene Aliases
DPC4, JIP, MADH4, MYHRS
Location
Chr.18:51030213-51085042 on build GRCh38
Assay Gene Location
Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
SMAD4 NM_005359.5 NP_005350.1
AK290770.1
BC002379.2 AAH02379.1
BX647129.1
KU178168.1
KU178169.1
U44378.1 AAA91041.1

View Full Product Details