Product Details

Assay Reference Genome
Location

Chr.1:20634476 on build GRCh38
Cytoband
1p36.12
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
65018
Gene Symbol
PINK1
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
Location
Chr.1:20633455-20651511 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PINK1 NM_032409.2 NP_115785.1
AB053323.1 BAB55647.1
AF316873.1 AAK28062.1
BC028215.1 AAH28215.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv545731 Chr1:19989106 - 20776306 on Build GRCh38 Gain UBXN10 SH2D5 LINC01141 HP1BP3 CDA FAM43B VWA5B1 CAMK2N1 PINK1-AS KIF17 MIR6084 PLA2G5 DDOST PLA2G2F PLA2G2C MUL1 PLA2G2D UBXN10-AS1 PINK1
nsv834369 Chr1:20609770 - 20779837 on Build GRCh38 Loss KIF17 MIR6084 SH2D5 DDOST HP1BP3 CDA PINK1-AS PINK1

View Full Product Details