Product Details

Assay Reference Genome
Location

Chr.Y:12719322 on build GRCh38
Cytoband
Yq11.221
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8287
Gene Symbol
USP9Y
Gene Name
ubiquitin specific peptidase 9, Y-linked
Gene Aliases
DFFRY, SPGFY2
Location
Chr.Y:12701231-12860844 on build GRCh38
Assay Gene Location
Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
USP9Y NM_004654.3 NP_004645.2
XM_017030078.1 XP_016885567.1
AF000986.2 AAC51833.1
Y13618.1 CAA73940.1
Y13619.1 CAA73941.1

View Full Product Details