Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193376.1 | NP_001180305.1 | 3 - 4 | 1824 | 94 |
NM_198253.2 | NP_937983.2 | 3 - 4 | 1824 | 94 | |
XM_017009796.1 | XP_016865285.1 | 3 - 4 | 2396 | 94 | |
GenBank mRNA | AB085628.1 | - | 3 - 4 | 1766 | 94 |
AB086379.1 | - | 3 - 4 | 1766 | 94 | |
AB086950.1 | - | 3 - 4 | 1766 | 94 | |
AF015950.1 | - | 3 - 4 | 1821 | 94 | |
AF018167.1 | - | 3 - 4 | 1822 | 94 | |
AX810036.1 | - | 3 - 4 | 1821 | 94 | |
BC062321.1 | - | 1 - 2 | 579 | 94 | |
JF896280.1 | - | 2 - 3 | 435 | 94 |