Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000594.3 | NP_000585.2 | 1 - 2 | 366 | 85 |
GenBank mRNA | AK314960.1 | - | 1 - 2 | 365 | 85 |
BC028148.1 | - | 1 - 2 | 348 | 85 | |
BI908079.1 | - | 1 - 2 | 361 | 86 | |
BP215875.1 | - | 1 - 2 | 366 | 85 | |
FJ795028.1 | - | 1 - 2 | 190 | 85 | |
HQ201306.2 | - | 1 - 2 | 191 | 85 | |
X01394.1 | - | 1 - 2 | 343 | 85 |