Gene Transcripts

Gene Symbol
Phrf1
Entrez Gene ID
101471
Gene Name
PHD and ring finger domains 1
Gene Aliases
AA673488
Chromosome Location
Chr. 7: 141228263 - 141262751 on Build GRCm38
UniGene ID
Mm.248656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081118.1 NP_001074587.1 8 874
GenBank mRNA BC138446.1 - 3 520
BC145719.1 - 3 520 40131022

View Full Product Details