Gene Transcripts

Gene Symbol
Ints4
Entrez Gene ID
101861
Gene Name
integrator complex subunit 4
Gene Aliases
2610034N24Rik, AA692684, AW493223
Chromosome Location
Chr. 7: 97480950 - 97541398 on Build GRCm38
UniGene ID
Mm.27383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027256.2 NP_081532.1 1 172
GenBank mRNA AK088635.1 - 1 116
AK163383.1 - 1 126
BC013710.1 - 1 118 3156446
CA450656.1 - 1 42

View Full Product Details